Skip to Content
Merck
All Photos(1)

Documents

8.22187

Sigma-Aldrich

TWEEN® 80

for synthesis

Synonym(s):

POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352104
EC Index Number:
500-019-9
NACRES:
NA.22

vapor pressure

<1.33 hPa

Quality Level

form

liquid

potency

42200 mg/kg LD50, oral (Rat)

pH

5-7 (20 °C, 50 g/L in H2O)

kinematic viscosity

300-500 cSt(25 °C)

bp

>100 °C/1013 hPa

transition temp

flash point >149 °C

density

1.07 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tween® 80 for synthesis. CAS 9005-65-6, pH 5 - 7 (50 g/l, H₂O, 20 °C).

Analysis Note

Density (d 20 °C/ 4 °C): 1.060 - 1.090
Saponification value: 45 - 55
Hydroxyl value: 65 - 80
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service