Skip to Content
Merck
All Photos(3)

Key Documents

8.40123

Sigma-Aldrich

Span® 80

greener alternative

for synthesis

Synonym(s):

Span® 80, Sorbitan monooleate, (Z)-Sorbitan mono-9-octadecenoate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$62.90
500 ML
$102.00
1 L
$184.00

$62.90


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$62.90
500 ML
$102.00
1 L
$184.00

About This Item

Empirical Formula (Hill Notation):
C24H44O6
CAS Number:
Molecular Weight:
428.60
MDL number:
UNSPSC Code:
12352108
EC Index Number:
215-665-4
NACRES:
NA.22

$62.90


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

<1.4 hPa ( 20 °C)

Quality Level

form

liquid

potency

>39800 mg/kg LD50, oral (Rat)

greener alternative product characteristics

Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

>100 °C

transition temp

flash point >149 °C

density

1.0 g/cm3 at 20 °C

greener alternative category

storage temp.

2-30°C

SMILES string

O1[C@@H]([C@H]([C@@H](C1)O)O)C(O)COC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C24H44O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22(27)29-19-21(26)24-23(28)20(25)18-30-24/h9-10,20-21,23-26,28H,2-8,11-19H2,1H3/b10-9-/t20-,21?,23+,24-/m1/s1

InChI key

NWGKJDSIEKMTRX-NZAQQJATSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is biodegradable surfactant, thus aligns with "Use of Renewable Feedstocks" and "Design for Degradation" principles of Green Chemistry, and belongs to "12 principles aligned" category of our greener alternatives.

Application

Span® 80 for synthesis. CAS 1338-43-8, molar mass 428.60 g/mol.

Analysis Note

Hydroxyl value: 193 - 209
Saponification value: 145 - 160
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Legal Information

Span is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service