Skip to Content
Merck
All Photos(1)

Key Documents

8.20584

Sigma-Aldrich

di-Sodium fumarate

for synthesis

Synonym(s):

di-Sodium fumarate, Fumaric acid disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$48.40
500 G
$114.00
1 KG
$197.00
25 KG
$2,240.00

$48.40


Estimated to ship on06 April 2025



Select a Size

Change View
100 G
$48.40
500 G
$114.00
1 KG
$197.00
25 KG
$2,240.00

About This Item

Linear Formula:
NaOOCCH=CHCOONa
CAS Number:
Molecular Weight:
160.04
MDL number:
UNSPSC Code:
12352107
EC Index Number:
241-087-7
NACRES:
NA.22

$48.40


Estimated to ship on06 April 2025


Quality Level

pH

7-8 (50 g/L in H2O)

solubility

228 g/L

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[O-]C(=O)\C=C/C(=O)[O-]

InChI

1S/C4H4O4.2Na/c5-3(6)1-2-4(7)8;;/h1-2H,(H,5,6)(H,7,8);;/q;2*+1/p-2/b2-1-;;

InChI key

MSJMDZAOKORVFC-UAIGNFCESA-L

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service