Skip to Content
Merck
All Photos(1)

Key Documents

8.00269

Sigma-Aldrich

Fumaric acid

for synthesis

Synonym(s):

Fumaric acid, trans-1,2-Ethylentricarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$31.60
1 KG
$66.00
2.5 KG
$133.00
50 KG
$1,230.00

$31.60


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$31.60
1 KG
$66.00
2.5 KG
$133.00
50 KG
$1,230.00

About This Item

Empirical Formula (Hill Notation):
C4H4O4
CAS Number:
Molecular Weight:
116.07
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-743-0
NACRES:
NA.22

$31.60


Estimated to ship on31 May 2025


Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

<0.001 hPa ( 20 °C)

Assay

≥99.0% (acidimetric)

form

solid

autoignition temp.

375 °C

potency

9300 mg/kg LD50, oral (Rat)
20000 mg/kg LD50, skin (Rabbit)

pH

2.1 (20 °C, 4.9 g/L in H2O)

bp

290 °C/1013 hPa (sublimed)

mp

287 °C

transition temp

flash point 273 °C

solubility

4.9 g/L

density

1.64 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

[O-]C(=O)\C=C\C(=O)[O-].[H+].[H+]

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+

InChI key

VZCYOOQTPOCHFL-OWOJBTEDSA-N

Related Categories

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

523.4 °F

Flash Point(C)

273 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service