Skip to Content
Merck
All Photos(2)

Key Documents

911364

Sigma-Aldrich

Poly(ethylene glycol) linear dendrimer

carboxyl terminated, PEG Mn 10k, generation 4

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$807.00

$807.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$807.00

About This Item

Empirical Formula (Hill Notation):
C732H1278O414
Molecular Weight:
16703.73
UNSPSC Code:
12162002
NACRES:
NA.23

$807.00


Check Cart for Availability

Request a Bulk Order

form

solid

mol wt

PEG Mn 10k
PEG average Mn 10,000
generation 4

no. Surface Groups

32

color

white to off-white

functional group

carboxylic acid

storage temp.

−20°C

General description

  • Polymer architecture: PEG-core dendrimer
  • Bis-MPA : 2,2-Bis(hydroxymethyl)propionic acid

Application

Bis-MPA dendritic molecules are based on 2,2-bismethylolpropionicacid (bis-MPA) building blocks. The hydrolyzable linker allows for degradation into byproducts that can be reabsorbed by the body or into excretable non-toxic small molecules. This dendritic polymer is biocompatible, biodegradable, and suitable for biomedical applications. PEG-core dendrimers feature excellent solubility in water and a high degree of functionality per PEG chain, enabling use for advanced drug delivery systems or for drug targeting. The carboxylic acid-functionalized corona can be readily used for bioconjugation reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of multifunctional polymers based on bis-MPA and their cutting-edge applications.
Garcia-Gallego S, et al.
Progress in Polymer Science, 48, 85-110 (2015)
Chemistry of multifunctional polymers based on bis-MPA and their cutting-edge applications.
Garcia-Gallego S, et al
Progress in Polymer Science, 48, 85-110 (2015)
Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Articles

Dendritic molecules' biomedical applications, particularly bis-MPA dendritic scaffolds, hold promise for drug delivery.

Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service