Skip to Content
Merck
All Photos(1)

Key Documents

8.14720

Sigma-Aldrich

Diisobutyldimethoxysilane

for synthesis

Synonym(s):

Diisobutyldimethoxysilane, Eurenor

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$95.00
100 ML
$303.00

$95.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
25 ML
$95.00
100 ML
$303.00

About This Item

Empirical Formula (Hill Notation):
C10H24O2Si
CAS Number:
Molecular Weight:
204.38
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

$95.00


0 left in stock

Request a Bulk Order

vapor pressure

0.75 hPa ( 25 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

255 °C

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

bp

188 °C/1025 hPa

mp

<-78 °C

transition temp

flash point 62 °C

density

0.863 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](OC)(OC)(CC(C)C)CC(C)C

InChI

1S/C10H24O2Si/c1-9(2)7-13(11-5,12-6)8-10(3)4/h9-10H,7-8H2,1-6H3

InChI key

NHYFIJRXGOQNFS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.860 - 0.865
Identity (IR): passes test

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

143.6 °F

Flash Point(C)

62 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service