Skip to Content
Merck
All Photos(1)

Key Documents

8.14719

Sigma-Aldrich

Diisopropyldimethoxysilane

for synthesis

Synonym(s):

Diisopropyldimethoxysilane, Eurenor 5021

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$344.00

$344.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
$344.00

About This Item

Empirical Formula (Hill Notation):
C8H20O2Si
CAS Number:
Molecular Weight:
176.33
MDL number:
UNSPSC Code:
12352103
EC Index Number:
421-540-7
NACRES:
NA.22

$344.00


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

2.14 hPa ( 20 °C)

Quality Level

Assay

≥95.0% (GC)

form

liquid

autoignition temp.

240 °C

potency

≥2000 mg/kg LD50, skin (Rat)

bp

164 °C/1013 hPa

mp

<-72 °C

transition temp

flash point 44 °C

density

0.88 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Si](OC)(OC)(C(C)C)C(C)C

InChI

1S/C8H20O2Si/c1-7(2)11(9-5,10-6)8(3)4/h7-8H,1-6H3

InChI key

VHPUZTHRFWIGAW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.877 - 0.882
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F

Flash Point(C)

44 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service