Skip to Content
Merck
All Photos(3)

Key Documents

260649

Sigma-Aldrich

Isoamyl ether

99%

Synonym(s):

Isopentyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$129.00
500 ML
$201.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$129.00
500 ML
$201.00

About This Item

Linear Formula:
(C5H11)2O
CAS Number:
Molecular Weight:
158.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$129.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.408 (lit.)

bp

173 °C (lit.)

solubility

alcohol: miscible(lit.)
chloroform: miscible(lit.)
diethyl ether: miscible(lit.)
water: insoluble(lit.)

density

0.778 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CC(C)CCOCCC(C)C

InChI

1S/C10H22O/c1-9(2)5-7-11-8-6-10(3)4/h9-10H,5-8H2,1-4H3

InChI key

AQZGPSLYZOOYQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isoamyl ether has been used in the synthesis of:
  • silver nanoparticles in a continuous flow tubular microreactor[1]
  • single-crystal Ag nanocubes (with slight truncation) of around 13.5nm in edge length[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of small silver nanocubes in a hydrophobic solvent by introducing oxidative etching with Fe (III) species.
Ma Y, et al.
Journal of Materials Chemistry, 20(18), 3586-3589 (2010)
Synthesis of silver nanoparticles in a continuous flow tubular microreactor.
Lin XZ, et al.
Nano Letters, 4(11), 2227-2232 (2004)
Martín E Villanueva et al.
Journal of colloid and interface science, 543, 247-255 (2019-03-02)
The interaction of hydrophobic silver nanoparticles with different phospholipids and stratum corneum mimic (SCM) membranes is studied in Langmuir monolayers. Thus, silver nanoparticles coated with oleic acid (AgNP-OA) were synthesized, characterized and incorporated in Langmuir monolayers of single phospholipids -having
Fuyao Liu et al.
Small (Weinheim an der Bergstrasse, Germany), 11(30), 3676-3685 (2015-04-29)
Hydrophobic ultrasmall nanoparticles synthesized in nonpolar solvents exhibit great potential in biomedical applications. However, a major challenge when applying these nanomaterials in biomedical research is the lack of a versatile strategy to render them water dispersible while preserving the hydrodynamic
Thiloka C Ariyasena et al.
Journal of chromatography. A, 1361, 240-254 (2014-08-30)
Retention factors on several columns and at various temperatures using gas chromatography and from reversed-phase liquid chromatography on a SunFire C18 column with various mobile phase compositions containing acetonitrile, methanol and tetrahydrofuran as strength adjusting solvents are combined with liquid-liquid

Articles

Ultrasonic spray pyrolysis produces scalable nanomaterials like metal oxides and quantum dots for diverse applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service