Skip to Content
Merck
All Photos(1)

Key Documents

SML2833

Sigma-Aldrich

JTE-607

≥98% (HPLC)

Synonym(s):

(-)-Ethyl-N-{3,5-dichloro-2-hydroxy-4-[2-(4-methylpiperazin-1-yl)ethoxy]benzoyl}-L-phenylalaninate dihydrochloride, JTE 607, N-[3,5-Dichloro-2-hydroxy-4-[2-(4-methyl-1-piperazinyl)ethoxy]benzoyl]-L-phenylalanine ethyl ester dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$246.00
25 MG
$790.00

$246.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$246.00
25 MG
$790.00

About This Item

Empirical Formula (Hill Notation):
C25H31Cl2N3O5·2HCl
CAS Number:
Molecular Weight:
597.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$246.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1c(c(cc(c1O)C(=O)N[C@@H](Cc3ccccc3)C(=O)OCC)Cl)OCCN2CCN(CC2)C.Cl.Cl

InChI

1S/C25H31Cl2N3O5.2ClH/c1-3-34-25(33)20(15-17-7-5-4-6-8-17)28-24(32)18-16-19(26)23(21(27)22(18)31)35-14-13-30-11-9-29(2)10-12-30;;/h4-8,16,20,31H,3,9-15H2,1-2H3,(H,28,32);2*1H/t20-;;/m0../s1

InChI key

JUJAUEQJEWIWCQ-FJSYBICCSA-N

Biochem/physiol Actions

JTE-607 is a pro-drug, which carboxylic form (JTE-607-COOH) is a potent cytokine release inhibitor. JTE-607 reduces the production of proinflammatory cytokines in models of acute injury, septic shock and endotoxemia. It also inhibits proliferation of AML cell in vitro and in xenograft model. JTE-607 is an inhibitor of CPSF3 (pre-messenger RNA endonuclease cleavage and polyadenylation specificity factor 3) that directly binds to CPSF3 and blocks the release of newly synthesized pre-mRNAs.
pro-drug of cytokine release inhibitor JTE-607-COOH; inhibitor of CPSF3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuyuki Tajima et al.
Cancer science, 101(3), 774-781 (2009-12-24)
Proinflammatory cytokines and growth factors have been thought to play crucial roles in the pathology of acute myelogenous leukemia (AML) by supporting the proliferation and survival of AML cells in an autocrine and paracrine manner, although further elucidation is required.
Junya Kakegawa et al.
Biochemical and biophysical research communications, 518(1), 32-37 (2019-08-11)
JTE-607 is a small molecule that was developed as an inflammatory cytokine inhibitor and also as an anti-leukemia reagent for monocytic leukemia. However, the mode of action of JTE-607 remains unknown. In this study, we identified JTE-607 to be a
Nathan T Ross et al.
Nature chemical biology, 16(1), 50-59 (2019-12-11)
The post-genomic era has seen many advances in our understanding of cancer pathways, yet resistance and tumor heterogeneity necessitate multiple approaches to target even monogenic tumors. Here, we combine phenotypic screening with chemical genetics to identify pre-messenger RNA endonuclease cleavage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service