Skip to Content
Merck
All Photos(1)

Key Documents

PZ0327

Sigma-Aldrich

PF06650833

≥98% (HPLC)

Synonym(s):

1-[[(2S,3S,4S)-3-Ethyl-4-fluoro-5-oxo-2-pyrrolidinyl]methoxy]-7-methoxy-6-isoquinolinecarboxamide, PF-06650833

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$208.00

$208.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$208.00

About This Item

Empirical Formula (Hill Notation):
C18H20FN3O4
CAS Number:
Molecular Weight:
361.37
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$208.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

NC(C1=C(OC)C=C2C(C=CN=C2OC[C@@H]3[C@H](CC)[C@H](F)C(N3)=O)=C1)=O

InChI

1S/C18H20FN3O4/c1-3-10-13(22-17(24)15(10)19)8-26-18-11-7-14(25-2)12(16(20)23)6-9(11)4-5-21-18/h4-7,10,13,15H,3,8H2,1-2H3,(H2,20,23)(H,22,24)/t10-,13+,15-/m0/s1

InChI key

JKDGKIBAOAFRPJ-ZBINZKHDSA-N

Biochem/physiol Actions

PF06650833 has been studied for its use in the treatment of Waldenstrom macroglobulinemia, indicated by overproduction of IgM (immunoglobulin M)-producing lymphoplasmacytic cells.[1]
PF06650833 is an inhibitor of Interleukin-1 receptor associated kinase 4 (IRAK4). IRAK4 kinase is important in innate immunity, involved in initiating signaling from Toll-like receptors and members of th einterleukin-1 receptor family. IRAK4 kinase is an attractive target for the treatment of various diseases associated with deregulated inflammation, such as rheumatoid arthritis, osteoarthritis, inflammatory bowel disease, and systemic lupus erythematosus. PF06650833 has been investigated for treatment of lupus.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sadiq Umar et al.
Cellular & molecular immunology (2020-05-18)
Flares of joint inflammation and resistance to currently available biologic therapeutics in rheumatoid arthritis (RA) patients could reflect activation of innate immune mechanisms. Herein, we show that a TLR7 GU-rich endogenous ligand, miR-Let7b, potentiates synovitis by amplifying RA monocyte and
Rafael Deliz-Aguirre et al.
The Journal of cell biology, 220(7) (2021-05-07)
A recurring feature of innate immune receptor signaling is the self-assembly of signaling proteins into oligomeric complexes. The Myddosome is an oligomeric complex that is required to transmit inflammatory signals from TLR/IL1Rs and consists of MyD88 and IRAK family kinases.
Kinase inhibitors in clinical practice: An expanding world.
Pandey R and Reuben K
The Journal of Allergy and Clinical Immunology, 141(2), 522-524 (2018)
Future therapeutic options for patients with Waldenstrom macroglobulinemia.
Castillo J J, et al.
Best Practice & Research. Clinical Haematology, 29(2), 206-215 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service