Skip to Content
Merck
All Photos(2)

Key Documents

524748

Sigma-Aldrich

PDE4D Inhibitor, GEBR-7b

The PDE4D Inhibitor, GEBR-7b controls the biological activity of PDE4D. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

PDE4D Inhibitor, GEBR-7b, GEBR-7b, (E)-3-(cyclopentyloxy)-4-methoxybenzaldehyde, O-2-(2,6-dimethylmorpholino)-2-oxoethyl oxime

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$281.00

$281.00


Estimated to ship on22 May 2025



Select a Size

Change View
5 MG
$281.00

About This Item

Empirical Formula (Hill Notation):
C21H30N2O5
CAS Number:
Molecular Weight:
390.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$281.00


Estimated to ship on22 May 2025


Quality Level

Assay

≥95% (HPLC)

form

semisolid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 100 mg/mL, clear, colorless

shipped in

wet ice

storage temp.

−20°C

SMILES string

COC1=C(OC2CCCC2)C=C(/C([H])=N/OCC(N3CC(C)OC(C)C3)=O)C=C1

InChI key

JEXHQZPMUKFOIV-SSDVNMTOSA-N

General description

A cell permeable, potent, and selective oxime PDE4D inhibitor (IC50 = 0.67 µM) which displays much reduced activities toward other PDE4 isoforms. It is shown to cause a significant 40% increase in extracellular cAMP in the hippocampus of freely moving rats in vivo at 100 µM, but has no effect on Aβ levels. This compound elicits cognitive enhancement in vivo at the optimum dose of 0.003 mg/kg in both mice and rat models, and unlike rolipram, does not cause emesis-like behavior in rodents.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Bruno, O., et al. 2011. Br. J. Pharmacol.in press.
Bruno, O., et al. 2009. J. Med. Chem.52, 6546.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service