Skip to Content
Merck
All Photos(1)

Documents

344282

Sigma-Aldrich

Forskolin

from Coleus forskohlii, ≥97% (HPLC), liquid, positive inotropic agent, Calbiochem

Synonym(s):

Forskolin, Coleus forskohlii in DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O7
Molecular Weight:
410.50
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Forskolin, Coleus forskohlii in DMSO, The major cell-permeable diterpene isolated from the Indian plant Coleus forskohlii.

Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

The major cell-permeable diterpene isolated from the Indian plant Coleus forskohlii.
The major cell-permeable diterpene isolated from the Indian plant Coleus forskohlii. At low doses, it acts as a positive inotropic agent. At higher doses, it serves as a hypotensive and vasodilatory agent due to its actions as a smooth muscle relaxant. No major side effects are observed at effective doses. Forskolins pharmacological activities are due to its activation of adenylate cyclase (EC50 = 4 µM), resulting in increased cAMP levels. The exact mechanism of forskolins positive inotropic effect is unknown but may be related to a cAMP-dependent increase in Na+ permeability that results in indirect augmentation of Ca2+ release. Inhibits MAP kinase in rat renal mesangial cells (IC50 = 25 µM). Also acts as a Hh pathway antagonist. Shown to inhibit apoptosis in cerebellar granule cells and to induce apoptosis in resting human B lymphocytes.
The major cell-permeable diterpene isolated from the Indian plant Coleus forskohlii. At low doses, it acts as a positive inotropic agent. At higher doses, it serves as a hypotensive and vasodilatory agent due to its actions as a smooth muscle relaxant. No major side effects are observed at effective doses. Forskolins pharmacological activities are due to its activation of adenylate cyclase (EC50 = 4 µM), resulting in increased cAMP levels. The exact mechanism of forskolins positive inotropic effect is unknown but may be related to a cAMP-dependent increase in Na+ permeability that results in indirect augmentation of Ca2+ release. Inhibits MAP kinase in rat renal mesangial cells (IC50 = 25 µM). Also acts as a Hh pathway antagonist. Shown to inhibit apoptosis in cerebellar granule cells and to induce apoptosis in resting human B lymphocytes. The solid form of this compound (Cat. No. 344270) is also available.

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 50 mM (5 mg/244 µl) solution of Forskolin, Coleus forskohlii (Cat. No. 344270) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

D′Orazio, J.A., et al. 2006. Nature443, 340.
Noveen, A., et al. 1996. Biochem. Biophys. Res. Commun.219, 180.
Galli, C., et al. 1995. J. Neurosci.15, 1172.
Li, X., et al. 1995. Am. J. Physiol.269, C986.
Lomo, J., et al. 1995. J. Immunol.154, 1634.
Uneyama, H., et al. 1993. J. Biol. Chem.268, 168.
Laurenza, A., et al. 1989. Trends Pharmacol. Sci.10, 442.
Adashi, E.Y., and Resnick, C.E. 1986. J. Cell. Biochem.31, 217.
Seamon, K.B., and Daly, J.W. 1986. Adv. Cyclic Nucleotide Protein Phosphorylation Res.20, 1.
Huang, R., et al. 1982. Cyclic Nucleotide Res.8, 385.
Metzger, H., and Lindner, E. 1981. IRCS Med. Sci. Biochem. Cardiovasc. System Pharmacol.9, 99.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service