Skip to Content
Merck
All Photos(1)

Key Documents

557502

Sigma-Aldrich

Ro-20-1724

≥98% (TLC), solid, cAMP phosphodiesterase inhibitor, Calbiochem

Synonym(s):

Ro-20-1724, 4-(3-Butoxy-4-methoxybenzyl)-2-imidazolidinone

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$176.00

$176.00


Estimated to ship on02 June 2025



Select a Size

Change View
100 MG
$176.00

About This Item

Empirical Formula (Hill Notation):
C15H22N2O3
CAS Number:
Molecular Weight:
278.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$176.00


Estimated to ship on02 June 2025


Product Name

Ro-20-1724, A cell-permeable, selective inhibitor of cAMP-specific phosphodiesterase (PDE IV; IC₅₀ = 2 µM).

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 20%
ethanol: 4%

shipped in

ambient

storage temp.

10-30°C

SMILES string

N1C(CNC1=O)Cc2cc(c(cc2)OC)OCCCC

InChI

1S/C15H22N2O3/c1-3-4-7-20-14-9-11(5-6-13(14)19-2)8-12-10-16-15(18)17-12/h5-6,9,12H,3-4,7-8,10H2,1-2H3,(H2,16,17,18)

InChI key

PDMUULPVBYQBBK-UHFFFAOYSA-N

General description

A cell-permeable, selective inhibitor of cAMP-specific phosphodiesterase (PDE IV; IC50 = 2 µM). Inhibits superoxide generation and arachidonic-induced platelet aggregation. Also inhibits fMLP-induced neutrophil adhesion to vascular endothelial cells.
Selective inhibitor of cAMP-specific phosphodiesterase (PDE IV, IC50 = 2 µM). Inhibits superoxide generation and arachidonic-induced platelet aggregation. Also inhibits fMLP-induced neutrophil adhesion to vascular endothelial cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PDE IV
Product does not compete with ATP.
Reversible: no
Target IC50: 2 µM against PDE IV

Warning

Toxicity: Irritant (B)

Other Notes

Derian, C.K., et al. 1995. J. Immunol. 154, 308.
Rubin, L.L., et al. 1991. J. Cell Biol.115, 1725.
Lad, P.M., et al. 1985. Biochim. Biophys. Acta 846, 286.
Bergstrand, H., et al. 1977. Mol. Pharmacol.13, 38.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Keyvanloo Shahrestanaki et al.
Research in pharmaceutical sciences, 14(2), 107-114 (2019-10-18)
Diabetes incidence showed ascending trends in recent years indicating urgent need for new therapeutic agents. Extracellular adenosine signaling showed promising results. However, role of its A3 receptor in pancreatic β-cells proliferation and insulin secretion is not well established. Thus, we
Pieter J Gaillard et al.
Methods in molecular biology (Clifton, N.J.), 437, 161-175 (2008-03-29)
Drug delivery across the blood-brain barrier (BBB) is a major obstacle for the development of effective treatments of many central nervous system disorders. Sophisticated cell culture models of the BBB have helped us to identify, characterize, and validate a novel
Alejandro Sánchez-Melgar et al.
Frontiers in neuroscience, 15, 702817-702817 (2021-09-21)
Gliomas are the most common and aggressive primary tumors in the central nervous system. The nucleoside adenosine is considered to be one major constituent within the tumor microenvironment. The adenosine level mainly depends on two enzymatic activities: 5'-nucleotidase (5'NT or
Jeff A Beeler et al.
Methods in molecular biology (Clifton, N.J.), 237, 39-53 (2003-09-23)
This chapter outlines methods to purify soluble adenylyl cyclase (AC7) expressed in an Escherichia coli (E. coli) heterologous expression system. Guidelines are provided for constructing the expression plasmids, optimizing expression, culturing, and purifying the proteins. Purification requires two chromatographic steps.
Moumita Sarkar et al.
iScience, 25(7), 104527-104527 (2022-06-28)
The post-menopausal state in women is associated with increased cancer incidence, the reasons for which remain obscure. Curiously, increased circulating levels of beta-hCG (human chorionic gonadotropin) (a hormonal subunit linked with tumors of several lineages) are also often observed post-menopause.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service