Skip to Content
Merck
All Photos(1)

Key Documents

513033

Sigma-Aldrich

PD 168393

A potent, cell-permeable, irreversible, ATP-competitive and selective inhibitor of EGF receptor (EGFR) tyrosine kinase activity (IC₅₀ = 700 pM).

Synonym(s):

PD 168393, 4-[(3-Bromophenyl)amino]-6-acrylamidoquinazoline

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$322.00

$322.00


Estimated to ship on25 May 2025



Select a Size

Change View
1 MG
$322.00

About This Item

Empirical Formula (Hill Notation):
C17H13BrN4O
CAS Number:
Molecular Weight:
369.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$322.00


Estimated to ship on25 May 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Brc1cc(ccc1)Nc2ncnc3c2cc(cc3)NC(=O)C=C

InChI

1S/C17H13BrN4O/c1-2-16(23)21-13-6-7-15-14(9-13)17(20-10-19-15)22-12-5-3-4-11(18)8-12/h2-10H,1H2,(H,21,23)(H,19,20,22)

InChI key

HTUBKQUPEREOGA-UHFFFAOYSA-N

General description

A potent, cell-permeable, irreversible, ATP-competitive and selective inhibitor of EGF receptor (EGFR) tyrosine kinase activity (IC50 = 0.70 nM). Does not inhibit other protein kinases. Acts by binding to the catalytic domain of the EGFR with a 1:1 stoichiometry and alkylating Cys773. Excellent antitumor agent in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
EGFR
Product competes with ATP.
Reversible: no
Target IC50: 700 pM against EGFR

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Avoid freeze/thaw cycles of solutions.

Other Notes

Fry, D.W., et al. 1998. Proc. Natl. Acad. Sci USA 95, 12022.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service