Skip to Content
Merck
All Photos(1)

Key Documents

324518

Sigma-Aldrich

eIF4E/eIF4G Interaction Inhibitor II, 4E1RCat

The eIF4E/eIF4G Interaction Inhibitor II, 4E1RCat controls the biological activity of eIF4E/eIF4G interaction. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

eIF4E/eIF4G Interaction Inhibitor II, 4E1RCat, 4-((3 E)-3-((5-(4-Nitrophenyl)furan-2-yl)methylidene)-2-oxo-5-phenyl-2,3-dihydro-1H-pyrrol-1-yl)benzoic acid, eIF4F Inhibitor II

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$303.00

$303.00


Estimated to ship on22 May 2025



Select a Size

Change View
10 MG
$303.00

About This Item

Empirical Formula (Hill Notation):
C28H18N2O6
Molecular Weight:
478.45
UNSPSC Code:
12352200
NACRES:
NA.32

$303.00


Estimated to ship on22 May 2025


Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark brown

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable oxopyrrolyl benzoate compound that blocks the assembly of eukaryotic translation initiation factor (eIF) 4F complex (eIF4F) as well as disrupts preformed eIF4F by competing against eIF4G for eIF4E binding. Specifically blocks cap-dependent, but not cap-independent, protein translations in cell-free translation assays and inhibits protein, but not DNA or RNA, synthesis in MDA-MB-231 and HeLa cultures (50 µM) in a reversible manner. Although not effective when administered alone (15 mg/kg, i.p. daily for 5 days), 4E1RCat is shown to significantly prolong Pten+/-Eµ-myc and Tsc+/-Eµ-myc, but not Eµ-myc, tumor remission time when combined with Doxorubicin (Cat. No. 324380) treatment (10 mg/kg, i.p. once on day 2) in mice in vivo. Unlike 4EGI-1 (Cat. No. 324517), 4E1RCat also blocks eIF4E and 4E-BP1 interaction.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Very fluffy material, not recommended for subdividing solid material by hand-weighing. Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Cencic, R., et al. 2011. Proc. Natl. Acad. Sci. USA108, 1046.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service