Skip to Content
Merck
All Photos(1)

Key Documents

SML0506

Sigma-Aldrich

LY-411575

≥97% (HPLC)

Synonym(s):

LSN-411575, LY 411575, N2-[(2S)-2-(3,5-Difluorophenyl)-2-hydroxyethanoyl]-N1-[(7S)-5-methyl-6-oxo-6,7-dihydro-5H-dibenzo[b,d]azepin-7-yl]-L-alaninamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$169.00
25 MG
$673.00

$169.00


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$169.00
25 MG
$673.00

About This Item

Empirical Formula (Hill Notation):
C26H23F2N3O4
CAS Number:
Molecular Weight:
479.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$169.00


Available to ship on16 April 2025Details


Request a Bulk Order

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)[C@@H](O)c1cc(F)cc(F)c1)C(=O)N[C@@H]2C(=O)N(C)c3ccccc3-c4ccccc24

InChI

1S/C26H23F2N3O4/c1-14(29-25(34)23(32)15-11-16(27)13-17(28)12-15)24(33)30-22-20-9-4-3-7-18(20)19-8-5-6-10-21(19)31(2)26(22)35/h3-14,22-23,32H,1-2H3,(H,29,34)(H,30,33)/t14-,22-,23-/m0/s1

InChI key

ULSSJYNJIZWPSB-CVRXJBIPSA-N

Application

LY-411575 has been used as a potent γ-secretase inhibitor and Notch inhibitor.[1][2][3]

Biochem/physiol Actions

LY-411575 is potent, cell permeable and selective γ-secretase inhibitor that reduces Aβ/42 after acute or chronic treatment, and blocks Notch activation. Studies (Cancer Cell) have shown that treatment with LSN-411575 arrests KrasG12V-driven NSCLCs in association with inhibition of HES1 expression and ERK phosphorylation.
LY-411575 is potent, cell permeable and selective γ-secretase inhibitor that reduces Aβ40/42 after acute or chronic treatment, and blocks Notch activation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic assays for studying intramembrane proteolysis.
Bolduc D M, et al.
Methods in Enzymology, 584, 295-308 (2017)
K Pandya et al.
British journal of cancer, 105(6), 796-806 (2011-08-19)
We reported that Notch-1, a potent breast oncogene, is activated in response to trastuzumab and contributes to trastuzumab resistance in vitro. We sought to determine the preclinical benefit of combining a Notch inhibitor (γ-secretase inhibitor (GSI)) and trastuzumab in both
Different developmental histories of beta-cells generate functional and proliferative heterogeneity during islet growth.
Singh S P, et al.
Nature Communications, 8(1), 664-664 (2017)
Kyung-Won Park et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 29(7), 2294-2307 (2021-03-02)
Numerous aggregation inhibitors have been developed with the goal of blocking or reversing toxic amyloid formation in vivo. Previous studies have used short peptide inhibitors targeting different amyloid β (Aβ) amyloidogenic regions to prevent aggregation. Despite the specificity that can be
Prion-protein-interacting amyloid-β oligomers of high molecular weight are tightly correlated with memory impairment in multiple Alzheimer mouse models.
Kostylev M A, et al.
The Journal of Biological Chemistry, 290(28), 17415-17438 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service