Skip to Content
Merck
All Photos(1)

Key Documents

5097R

Sigma-Aldrich

CMV pp65 MONOCLONAL ANTIBODY

Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$1,110.00

$1,110.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$1,110.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.41

$1,110.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgG

shipped in

wet ice

General description

One dropper vial containing 5 mL of a blend of monoclonal antibodies specific for cytomegalovirus (CMV) pp65 antigen in buffer containing protein stabilizer, and 0.1% sodium azide (preservative). A replacement component for CHEMICON Light Diagnostics Catalog No. 3244 and 3247
Ready to use

Specificity

Specific for cytomegalovirus (CMV) pp65 antigen.

Application

Detect CMV pp65 MONOCLONAL using this CMV pp65 monoclonal antibody validated for use in immunofluorescence.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
One dropper vial containing 5 mL of a blend of monoclonal antibodies specific for cytomegalovirus (CMV) pp65 antigen in buffer containing protein stabilizer, and 0.1% sodium azide (preservative).
Protein A purified

Storage and Stability

When stored at 2° to 8°C, this reagent is stable up to the expiration date printed on the label. Do not freeze or expose to elevated temperatures. Discard any remaining reagent after the expiration date.

Analysis Note

Control
CMV infected BAL Patient Sample

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service