Skip to Content
Merck
All Photos(1)

Documents

MAB8121

Sigma-Aldrich

Anti-Cytomegalovirus Antibody, blend, clone 8B1.2, 1G5.2, and 2D4.2

Chemicon®, from mouse

Synonym(s):

CMV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G5.2, monoclonal
2D4.2, monoclonal
8B1.2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Reacts with Immediate early, early, and late antigen preparations. Can detect CMV infection within 2 hours post-infection exhibiting a nuclear staining which reaches peak intensity between 48 and 96 hours. This antigen is persisting and can be detected during the complete CMV infection cycle. Staining of CMV structural antigens becomes apparent at about 48 hours. Extra-nuclear staining is first seen in the region of the golgi apparatus followed subsequently by staining of viral proteins and particles in the cytoplasm.

With CMV the antigens expressed at different times are listed as:



Immediate Early (alpha gene expression): those antigens expressed at 3-12 hrs post-infection generally involved in Transcription such as 72kD major phosphoprotein and a few other other antigens at 60 - 80kD.

Early Antigen (beta genes) a.k.a. Delayed Early or Intermediate Early: expressed at 12-24hrs post-infection. Generally enzymes and one virion structural gene preceding viral DNA synthesis.

Late Antigen (gamma genes): expressed at 36-48hrs post-infection. Generally structural proteins. Major protein = 55kD

Immunogen

Epitope: blend
MRC-5 infected cells with ATCC VR538 and AD169, ultrasonicated, screened by plate reduction neutralization tests.

Application

Immunohistochemistry

Immunofluorescence

Optimal working dilutions must be determined by the end user.
This Anti-Cytomegalovirus Antibody, blend, clone 8B1.2, 1G5.2 & 2D4.2 is validated for use in IF, IH for the detection of Cytomegalovirus.

Physical form

0.02 M PB, 0.25M NaCl, PH 7.6 with 0.1% Sodium Azide.
Format: Purified

Storage and Stability

Maintain at +2-8C in convenient aliquots for up to 6 months. Do not freeze.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intrauterine growth restriction caused by underlying congenital cytomegalovirus infection.
Pereira, L; Petitt, M; Fong, A; Tsuge, M; Tabata, T; Fang-Hoover, J; Maidji, E; Zydek et al.
The Journal of Infectious Diseases null
Takako Tabata et al.
The American journal of pathology, 186(11), 2970-2986 (2016-10-30)
Human cytomegalovirus (HCMV) is the leading viral cause of birth defects, including microcephaly, neurological deficits, hearing impairment, and vision loss. We previously reported that epithelial cells in amniotic membranes of placentas from newborns with intrauterine growth restriction and underlying congenital
Takako Tabata et al.
Journal of virology, 89(9), 5134-5147 (2015-03-06)
Human cytomegalovirus (HCMV) is a major cause of birth defects that include severe neurological deficits, hearing and vision loss, and intrauterine growth restriction. Viral infection of the placenta leads to development of avascular villi, edema, and hypoxia associated with symptomatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service