Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200678

Sigma-Aldrich

Anti-Collagen Type I antibody, Mouse monoclonal

clone COL-1, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Collagen Type I antibody produced in mouse, OI4, alpha 1, alpha 2, collagen, type I

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$830.00

$830.00


Estimated to ship on03 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$830.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

$830.00


Estimated to ship on03 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

COL-1, monoclonal

form

buffered aqueous solution

species reactivity

pig, rat, human, deer, bovine

concentration

~1 mg/mL

technique(s)

ELISA: suitable
dot blot: suitable
immunoblotting: 1-2 μg/mL using recombinant human collagen, expressed in Nicotiana tabacum
immunohistochemistry: 3.5-7 μg/mL using frozen sections of human tonsil, human tongue or pig tongue

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL1A1(1277)

General description

Collagen type Iα1 (COL1A1) is encoded by the gene that is located on human chromosome 17q21.33. It is the most abundant extracellular matrix (ECM) protein in humans. Type 1 collagen is the major structural protein of bone, tendon, skin and cornea. The encoded protein is a heterotrimer consisting of two α1-chains and one α2-chain.
Collagens are present in the extracellular matrix and contains a chains which can differ in length.

Immunogen

bovine skin collagen type I

Application

Monoclonal Anti-Collagen Type I antibody produced in mouse has been used in immunohistochemical analysis and in vitro invasion assays.

Biochem/physiol Actions

Collagen, type I (COL1A1) is an important component of the connective tissue matrix. It plays an important role in the growth and maintenance of organ and tissue integrity. This protein also participates in the process of tissue repair. COL1A1 is implicated in fibrosis.
Collagens are involved in structural roles. Additionally, they associate with various receptors and control cell proliferation, migration as well as differentiation.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Antonia Martínez-Pascual et al.
International journal of molecular sciences, 25(14) (2024-07-27)
Androgenic alopecia (AGA) is the most common type of alopecia and its treatments involve drugs that have various adverse effects and are not completely effective. Radiofrequency-based therapies (RF) are an alternative for AGA treatment. Although there is increasing clinical evidence
Paeoniflorin prevents postoperative peritoneal adhesion formation in an experimental rat model.
Gao Q, et al.
Oncotarget, 8(55), 93899-93899 (2017)
Deregulation of the platelet-derived growth factor β-chain gene via fusion with collagen gene COL1A1 in dermatof ibrosarcoma protuberans and giant-cell fibroblastoma.
Simon M-P, et al.
Nature Genetics, 15(10), 95-95 (1997)
Overexpression of CXCR4 promotes invasion and migration of non?small cell lung cancer via EGFR and MMP?9.
Zuo J, et al.
Oncology Letters, 14(6), 7513-7521 (2017)
Bruna B Catinelli et al.
Scientific reports, 12(1), 7375-7375 (2022-05-06)
Gestational diabetes mellitus (GDM) plus rectus abdominis muscle (RAM) myopathy predicts long-term urinary incontinence (UI). Atrophic and stiff RAM are characteristics of diabetes-induced myopathy (DiM) in pregnant rats. This study aimed to determine whether swimming exercise (SE) has a therapeutic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service