Skip to Content
Merck
All Photos(1)

Documents

5.08314

Sigma-Aldrich

KN-92

InSolution, ≥95%, 5 mM in DMSO, Useful as a negative control for KN-93

Synonym(s):

InSolution KN-92, 2-(N-(4-Methoxybenzenesulfonyl))amino-N-(4-chlorocinnamyl)-N-methylbenzylamine, Phosphate, ( E)-N-(2-(((3-(4-Chlorophenyl)allyl)(methyl)amino)methyl)phenyl)-4-methoxybenzenesulfonamide, Phosphate, 2-(N-(4-Methoxybenzenesulfonyl))amino-N-(4-chlorocinnamyl)-N-methylbenzylamine, Phosphate, (E)-N-(2-(((3-(4-Chlorophenyl)allyl)(methyl)amino)methyl)phenyl)-4-methoxybenzenesulfonamide, Phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25ClN2O3S · xH3PO4
CAS Number:
Molecular Weight:
456.98
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

InChI

1S/C24H25ClN2O3S.H3O4P/c1-27(17-5-6-19-9-11-21(25)12-10-19)18-20-7-3-4-8-24(20)26-31(28,29)23-15-13-22(30-2)14-16-23;1-5(2,3)4/h3-16,26H,17-18H2,1-2H3;(H3,1,2,3,4)/b6-5+;

InChI key

XRQHWVVDNMJDEQ-IPZCTEOASA-N

General description

Useful as a negative control for KN-93 (Cat. No. 422708), a Ca2+/CaM kinase II inhibitor.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 5 mM (500 µg/180 µL) sterile-filtered solution of KN-92 (Cat. No. 422709) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 3 months at -20°C.

Other Notes

Fan, G.H., et al. 1999. Mol. Pharmacol.56, 39.
Fan, G.H., et al. 1997. Neuropharmacology36, 1763.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service