Skip to Content
Merck
All Photos(1)

Documents

422712

Sigma-Aldrich

KN-93

InSolution, ≥95%, A cell-permeable, competitive inhibitor of Ca2+/calmodulin-dependent protein kinase II

Synonym(s):

InSolution KN-93

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29ClN2O4S
CAS Number:
Molecular Weight:
501.04
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

liquid

potency

370 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C26H29ClN2O4S/c1-28(17-5-6-21-9-11-23(27)12-10-21)20-22-7-3-4-8-26(22)29(18-19-30)34(31,32)25-15-13-24(33-2)14-16-25/h3-16,30H,17-20H2,1-2H3/b6-5+

InChI key

LLLQTDSSHZREGW-AATRIKPKSA-N

General description

A cell-permeable, competitive inhibitor of Ca2+/calmodulin-dependent protein kinase II (Ki = 370 nM). Selectively binds to the Calmodulin (CaM) binding site of the enzyme and prevents the association of CaM with CaM Kinase II. Inhibits glucose-induced and forskolin-stimulated insulin release from isolated pancreatic islets. Has no significant effect on protein kinase A activity. Induces G1 cell cycle arrest and apoptosis in NIH 3T3 cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
cam kinase 2
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 5 mM (1 mg/399 µl) solution of KN-93 (Cat. No. 422708) in DMSO.

Reconstitution

Following initial thaw aliquot and freeze (-20°C).

Other Notes

Fan, G.H., et al. 1999. Mol. Pharmacol.56, 39.
Tombes, R.M., et al. 1995. Cell Growth Differentiation6, 1063.
Chen, C., et al. 1994. Science266, 291.
Mamiya, N., et al. 1993. Biochem. Biophys. Res. Commun.195, 608.
Niki, I., et al. 1993. Biochem. Biophys. Res. Commun.191, 255.
Sumi, M., et al. 1991. Biochem. Biophys. Res. Commun.181, 968.
Tokumitsu, H., et al. 1990. J. Biol. Chem.265, 4315.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service