Skip to Content
Merck
All Photos(1)

Key Documents

PHR1158

Supelco

Benzocaine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

4-Aminobenzoic acid ethyl ester, Ethyl 4-aminobenzoate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$187.00

$187.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$187.00

About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
Beilstein:
638434
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$187.00


Available to ship on14 April 2025Details


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1054000

API family

benzocaine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCOC(=O)c1ccc(N)cc1

InChI

1S/C9H11NO2/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6H,2,10H2,1H3

InChI key

BLFLLBZGZJTVJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Benzocaine is an ester-type local anesthetic, able to induce pain relief. It is used extensively in topical, mucosal and dermal formulations.[1]

Application

Benzocaine may be used as a pharmaceutical reference standard for the determination of the analyte in bioadhesive gel,[1] pharmaceutical formulations[2] and channel catfish tissues[3] by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6506 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

233.6 °F

Flash Point(C)

112 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new validated method for the simultaneous determination of benzocaine, propylparaben and benzyl alcohol in a bioadhesive gel by HPLC.
Perez-Lozano P, et al.
Journal of Pharmaceutical and Biomedical Analysis, 39(5), 920-927 (2005)
Separation and simultaneous high-performance liquid chromatographic determination of benzocaine and benzyl benzoate in a pharmaceutical preparation.
Gigante B, et al.
Journal of Chromatography A, 549(5), 217-220 (1991)
A Szoke et al.
Journal of pharmaceutical and biomedical analysis, 16(1), 69-75 (1998-02-03)
Methods for extraction and gradient HPLC quantification were developed for benzocaine (BZ) and three of its metabolites to be used in conjunction with a reverse isotope technique. The metabolites were p-aminobenzoic acid (PABA), acetyl-p-aminobenzoic acid (AcPABA) and acetylbenzocaine (AcBZ). The
Qing Zhu et al.
Molecular pharmaceutics, 9(3), 546-553 (2012-02-04)
The crystallization kinetics of various active pharmaceutical ingredient/polyethylene glycol (API/PEG) solid dispersions has been investigated using wide-angle X-ray diffraction (XRD) and Raman spectroscopy. APIs with different physicochemical properties and crystallization tendency were employed to form solid dispersions with PEG. The
Nathalie Ferreira Silva De Melo et al.
Journal of pharmaceutical sciences, 101(3), 1157-1165 (2011-11-23)
This paper describes a comparison of different polymeric nanocapsules (NCs) prepared with the polymers poly(D,L-lactide-co-glycolide), poly(L-lactide) (PLA), and poly(ε-caprolactone) and used as carrier systems for the local anesthetic (LA) benzocaine (BZC). The systems were characterized and their anesthetic activities investigated.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service