Skip to Content
Merck
All Photos(1)

Key Documents

203781

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl-α-D-glucopyranoside

Synonym(s):

5-Bromo-4-chloro-3-indolyl-α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$342.00

$342.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$342.00

About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
MDL number:
UNSPSC Code:
41116133
NACRES:
NA.41

$342.00


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

2% DMF: soluble
DMSO: soluble

shipped in

ambient

storage temp.

−20°C

General description

Substrate for acid α-glucosidase, a glycogen-degrading lysosomal enzyme. Yields an intense blue color upon enzymatic cleavage.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service