Skip to Content
Merck
All Photos(1)

Key Documents

RES1364C

Sigma-Aldrich

Red-Gal®

Synonym(s):

6-Chloro-3-indolyl-β-D-galactopyranoside, Rose-Gal, Salmon-Gal

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$82.60
1 EACH
$133.00
1 EACH
$397.00

$82.60


Check Cart for Availability


Select a Size

Change View
1 EACH
$82.60
1 EACH
$133.00
1 EACH
$397.00

About This Item

Empirical Formula (Hill Notation):
C14H16ClNO6
CAS Number:
Molecular Weight:
329.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.21

$82.60


Check Cart for Availability

biological source

synthetic

Quality Level

form

powder

technique(s)

immunohistochemistry: suitable

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](On2ccc3ccc(Cl)cc23)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C14H16ClNO6/c15-8-2-1-7-3-4-16(9(7)5-8)22-14-13(20)12(19)11(18)10(6-17)21-14/h1-5,10-14,17-20H,6H2/t10-,11+,12+,13-,14+/m1/s1

InChI key

CCSMDIRWROWGMP-HTOAHKCRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Product is available in the following packages sizes:
RES1364C-A101X: 25 mg container
RES1364C-A102X: 100 mg container
RES1364C-A103X: 1 gm container

Legal Information

Red-Gal is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service