Skip to Content
Merck
All Photos(1)

Key Documents

XGAL-RO

Roche

X-Gal

solubility: in DMF, soluble, solubility: in methanol, soluble, storage temp.:2-8°C

Synonym(s):

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside, 5-bromo-4-chloro-3-indolyl-β-d-galactopyranoside, bromo-4-chloro-3-indolyl-β-d-galactopyranoside, 5-, 5-Bromo-4-chloro-3-indolyl β-D-galactoside, BCIG, X-Gal

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$186.00
250 MG
$350.00
1 G
$881.00

$186.00


Available to ship on10 April 2025Details



Select a Size

Change View
100 MG
$186.00
250 MG
$350.00
1 G
$881.00

About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
Beilstein:
1402009
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.28

$186.00


Available to ship on10 April 2025Details


description

5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside

Quality Level

Assay

99%

form

solid

mol wt

408.63 g/mol

packaging

pkg of 1 g (10745740001)
pkg of 100 mg (03117073001)
pkg of 100 mg (11680293001)
pkg of 2.5 g (10703729001)
pkg of 250 mg (10651745001)

manufacturer/tradename

Roche

concentration

20 mg/mL

technique(s)

activity assay: suitable

color

white

solubility

DMF: soluble
methanol: soluble

application(s)

genomic analysis
life science and biopharma

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C14H15BrClNO6/c15-5-1-2-6-9(10(5)16)7(3-17-6)22-14-13(21)12(20)11(19)8(4-18)23-14/h1-3,8,11-14,17-21H,4H2/t8-,11+,12+,13-,14-/m1/s1

InChI key

OPIFSICVWOWJMJ-AEOCFKNESA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

X-Gal is a chromogenic substrate for β-galactosidase that produces a rich blue color that can easily be detected visually over background. It is soluble in dimethylformamide and methanol. The product is not soluble in water.[1][2]

Application

X-Gal has been used for immunohistocytochemistry applications.[3] It is also the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.
X-Gal is a chromogenic substrate for β-galactosidase that produces a rich blue color that can easily be detected visually over background. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

Quality

Chromatographically homogeneous

Physical form

Crystals, white powder

Preparation Note

Working solution: Stock solution:
  • 20 mg/ml in dimethyl formamide (DMF), clear, colorless solution
  • A590 nm: 0.01

Storage conditions (working solution): X-gal in solution (i.e., dissolved in DMF 2% (w/v)) is stable for several weeks when stored in the dark at 2 to 8 °C; when the solution is stored at -15 to -25 °C, it is stable for one year.

Other Notes

For life science research only. Not for use in diagnostic procedures.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A I Geller et al.
Science (New York, N.Y.), 241(4873), 1667-1669 (1988-09-23)
A defective herpes simplex virus 1 (HSV-1) vector, pHSVlac, has been developed that contains a transcription unit that places the Escherichia coli lacZ gene under the control of the HSV-1 immediate early 4/5 promoter. The vector pHSVlac was propagated with
Maryvonne Arnaud et al.
Applied and environmental microbiology, 70(11), 6887-6891 (2004-11-06)
A shuttle vector designated pMAD was constructed for quickly generating gene inactivation mutants in naturally nontransformable gram-positive bacteria. This vector allows, on X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galactopyranoside) plates, a quick colorimetric blue-white discrimination of bacteria which have lost the plasmid, greatly facilitating clone
A R Yallowitz et al.
Cell death and differentiation, 21(4), 645-654 (2014-01-21)
In embryogenesis, p63 is essential to develop mammary glands. In the adult mammary gland, p63 is highly expressed in the basal cell layer that comprises myoepithelial and interspersed stem/progenitor cells, and has limited expression in luminal epithelial cells. In adult

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service