Skip to Content
Merck
All Photos(2)

Key Documents

146754

Millipore

Tryptic Soy Agar - Ready-to-use Contact Plates

IsoBag®, Lecithin, Sodium Thiosulfate, Tween®, Histidine, ICR with non-lockable lid, plate diam. 55 mm, 190 mm DPTE® alpha-port, sterile; irradiated, pack of 100 plates, suitable for surface monitoring

Synonym(s):

IsoBag® Tryptic Soy Agar Contact, Soybean Casein Digest Agar, TSA IsoBag®, TSA plate with neutralizers

Sign Into View Organizational & Contract Pricing

Select a Size

100 PKG
$2,420.00

$2,420.00


Estimated to ship on21 May 2025


Request more information

Select a Size

Change View
100 PKG
$2,420.00

About This Item

UNSPSC Code:
41106231
NACRES:
NA.74

$2,420.00


Estimated to ship on21 May 2025


Request more information

material

polystyrene plate

Quality Level

Agency

EP
USP

sterility

sterile; irradiated

form

plate
solid

feature

rapid transfer port
ready-to-use
190 mm DPTE® alpha-port

composition

Histidine
Lecithin
Sodium Thiosulfate
Tween®

packaging

pack of 100 plates

manufacturer/tradename

IsoBag®

plate diam.

55 mm

color

clear yellowish

pH

7.3 ( in H2O)

application(s)

pharmaceutical
surface monitoring

compatibility

stand alone

shipped in

ambient

storage temp.

15-25°C

suitability

bacteria
fungi

Looking for similar products? Visit Product Comparison Guide

General description

The IsoBag® TSA Contact is designed for quick and convenient transfer of culture media used for surface monitoring in isolators or RAB systems. The Beta bag consists of a sterile polyurethane DPTE Bag compatible with 190 mm Alpha Ports. The bag contains 55 mm contact plates filled with Tryptic Soy Agar.
TSA medium is used for the cultivation and isolation of a wide range of bacteria, yeasts, and molds. The formulation contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. Sodium chloride maintains the osmotic equilibrium, and the medium is supplemented with neutralizers such as lecithin, polysorbate (Tween®) 80, histidine, and sodium thiosulfate to inactivate disinfectant residues.

Application

The IsoBag® TSA Contact is designed for quick and convenient transfer of culture media needed for surface monitoring in isolators or RAB systems.

Legal Information

DPTE is a registered trademark of GETINGE LA CALHENE
ISOBAG is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

IsoBag® ease the environmental monitoring workflow in aseptic production isolator

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service