Skip to Content
Merck
All Photos(1)

Key Documents

1.46689

Millipore

ReadyPlate Chromocult® Coliform Agar

To detect Escherichia coli and coliform bacteria, plate diam. 90 mm, box of 20 plates

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$56.70

$56.70


In StockDetails


Request more information

Select a Size

Change View
20 EA
$56.70

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

$56.70


In StockDetails


Request more information

Agency

ISO 9308

Quality Level

sterility

sterile

form

plate (ready-to-use)

feature

ready-to-use

packaging

box of 20 plates

manufacturer/tradename

ReadyPlate

plate diam.

90 mm

application(s)

food and beverages
microbiology
water monitoring

storage temp.

15-25°C

suitability

Escherichia coli
coliforms

General description

ReadyPlate agar plates are stackable specially designed plates that helps increase the safety during handling, transport and incubation. ReadyPlate agar plates are of standard 90mm diameter. Chromocult® Coliform Agar (CCA) is a selective and differential chromogenic culture medium. It is a standard medium in water testing for simultaneous detection and enumeration of coliform and E.coli. It is also the base medium recommended for testing drinking water as per ISO 9309-1:2014

Application

Chromocult® Coliform Agar is used for the enumeration of Escherichia coli and coliform bacteria in food and drinking water, disinfected pool water and finished water from treatment plants

Analysis Note

The interaction of selected peptones, pyruvate, sorbitol and phosphate buffer promotes rapid colony growth, even for sub-lethally injured coliform bacteria. The growth of Gram-positive bacteria as well as certain Gram-negative bacteria is inhibited by the presence of Tergitol 7 which has no negative effect on the growth of coliform bacteria. Further, the combination of two chromogenic substrates permits the simultaneous detection of coliform bacteria and E. coli.
Coliform bacteria detection: The substrate Salmon-GAL is used for the detection of β-D-galactosidase activity, which is characteristic for coliform bacteria. This interaction results in a pink to red color of the coliform colonies.
E. coli detection: The substrate X-glucuronide is used for the detection of β-D-glucuronidase activity. E. coli cleaves both Salmon-GAL and X-glucuronide, so that positive colonies take on a dark-blue to violet color. These are easily distinguished from other coliform colonies, which have a pink to red color. Some E. coli (3-4%) are ß-D-glucuronidase-negative and appear as pink to red colonies.
Accompanying flora appears as colorless colonies, except for some organisms, which possess β-D glucuronidase activity. These colonies appear light blue to turquoise in color.

Legal Information

CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany
TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service