Skip to Content
Merck
All Photos(1)

Key Documents

1.02139

Millipore

Envirocheck Contact YM(R)

ready-to-use, for yeasts, for molds

Synonym(s):

Contact YM Microorganism Detection Kit, Contact Yeasts and Molds coated with CASO (Tryptic Soy) agar, Envirocheck Contact YM Yeasts and Molds, Envirocheck YM Testing Kit, Microbial Analysis Kit, Microbiological Detection Paddles, Tryptic Soy Agar Microbial Testing, Yeast and Mold Detection System

Sign Into View Organizational & Contract Pricing

Select a Size

10 UNITS
$44.30

$44.30


Available to ship on01 April 2025Details


Request more information

Select a Size

Change View
10 UNITS
$44.30

About This Item

UNSPSC Code:
41116116
NACRES:
NA.74

$44.30


Available to ship on01 April 2025Details


Request more information

material

plastic cap
plastic slide
plastic tube

Quality Level

product line

Envirocheck

form

solid

feature

ready-to-use

packaging

pack of 10 slides

manufacturer/tradename

Envirocheck

storage condition

(Tightly closed. Dry.)

application(s)

food and beverages
surface monitoring
water monitoring

storage temp.

2-30°C

suitability

molds
yeasts

General description

Envirocheck Contact Yeasts and Molds offer a semi-quantitative method to detect microorganisms on surfaces and in liquids. The flexible paddles are coated with Tryptic Soy Agar for yeast and mold analysis. Slide A uses CASO (Tryptic Soy) Agar with 0.05 % Triphenyl Tetrazolium chloride (TTC), where red colonies indicate bacterial growth through the formation of red formazan dye upon TTC reduction. Slide B is coated with Rose Bengal Chloramphenicol Agar, which inhibits bacterial growth while reducing the size and height of mold colonies, preventing overgrowth of slower-growing fungi by faster-growing species.

Application

Envirocheck Contact YM(R) slides can be used for the detection of yeasts and molds on bothsurface and liquid testing from environmental materials and foodstuffs. The contact slides are suitable for industrial and environmental monitoring.

Features and Benefits

  • Avoid freezing. Frozen agar shows a dimpled or fuzzy surface and cannot be used anymore.
  • Condensation can be prevented by avoiding quick temperature shifts and mechanical stress.
  • Upon storage, the dip slides should not be placed near heat sources such as refrigerators with heat-emitting condensers.

Analysis Note

Count the number of colonies on both sides of the paddle. CASO Agar with TTC: Count the number of colonies for obtaining the total aerobic bacterial count (TVC). Rose Bengal Chloramphenicol Agar: Count the number of colonies to obtain the fungal contamination (colonies of yeasts are red-pink)
Sterility: passes test
pH-value/side one (25 °C): 6.6 - 7.3
pH-value/side two (25 °C): 6.2 - 7.0
Growth/side one
- Escherichia coli ATCC 8739 (WDCM 00012):
good
- Staphylococcus aureus ATCC 6538 (WDCM 00032): fair/good
- Candida albicans ATCC 10231 (WDCM 00054): none/poor
- Saccharomyces cerevisiae ATCC 9763 (WDCM 00058): none/poor
- Pseudomonas aeruginosa ATCC 9027 (WDCM 00026): good
- Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053): good
Growth/side two
- Escherichia coli ATCC 8739 (WDCM 00012):
none
- Staphylococcus aureus ATCC 6538 (WDCM 00032): none
- Candida albicans ATCC 10231 (WDCM 00054): good
- Saccharomyces cerevisiae ATCC 9763 (WDCM 00058): good
- Pseudomonas aeruginosa ATCC 9027 (WDCM 00026): inhibited
- Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053): good
Incubation: 48 hrs.; 28 °C; aerobic; Aspergillus up to 72 hrs

Legal Information

ENVIROCHECK is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service