1.14661
Phosphate Test Kit, colorimetric
0.25-3.0 mg/L (PO₄³⁻), MQuant®
Synonym(s):
Phosphate Visual Test Kit
About This Item
Recommended Products
product name
Phosphate Test in freshwater and seawater, colorimetric, 0.25-3.0 mg/L (PO43-), for use with MQUANT®
specific analyte(s)
phosphate
Quality Level
measuring range
0.25-3.0 mg/L (PO43-)
compatibility
for use with MQUANT®
detection method
colorimetric
storage temp.
15-25°C
General description
Analysis Note
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service