Skip to Content
Merck
All Photos(1)

Key Documents

1.46000

Millipore

HEIMPLATE Violet Red Bile Dextrose Agar Settle plate

Heimplate, APHA, ISO 21528, USP, JP, plate diam. × filling volume 90 mm × 30 mL/ft

Synonym(s):

VRBD, Violet Red Bile Glucose agar, Crystal Violet Neutral Red Bile Glucose agar

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
$80.30

$80.30


Estimated to ship on12 April 2025


Request more information

Select a Size

Change View
20 PKG
$80.30

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

$80.30


Estimated to ship on12 April 2025


Request more information

Agency

APHA
EP
ISO 21528
JP
USP

Quality Level

sterility

sterile

form

plate (ready-to-use)

feature

ready-to-use
ICR Settle plate

manufacturer/tradename

Heimplate

packaging

pkg of (20 or 120 plates per box)

technique(s)

microbiological culture: suitable

plate diam. × filling volume

90 mm × 30 mL/ft

pH

7.4 ( in H2O)

application(s)

microbiology
pharmaceutical
surface monitoring

storage temp.

15-25°C

suitability

Escherichia coli
Salmonella spp.
coliforms

General description

Violet Red Bile Dextrose (VRBD) Agar settle plates are used for the detection and enumeration of Enterobacteriaceae and bile-tolerant Gram-negative bacteria. Crystal violet and bile salts inhibit the accompanying bacterial flora. Fermentation of glucose by Enterobacteriaceae leads to acid production, which is indicated by a color change to red and by zones of precipitated bile acids surrounding the colonies. Enzymatic digest of animal tissue provides carbon and nitrogen sources for the growth, and yeast extract primarily supplies the B-complex vitamins. The culture medium is not specific for Enterobacteriaceae as some other accompanying bacteria (e.g. Aeromonas) also show the same reaction.

Application

Violet Red Bile Dextrose (VRBD) Agar settle plates are used to detect and enumerate Enterobacteriaceae from food, animal feed, and other materials. They are also suitable for the enumeration of bile-tolerant Gram-negative bacteria from pharmaceuticals and other materials.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Protocols

The International Organization for Standardization (ISO) published the revised EN ISO 21528-1 and EN ISO 21528-2 standards in 2017.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service