Skip to Content
Merck
All Photos(1)

Key Documents

1.46048

Millipore

HEIMPLATE Cetrimide agar

HEIMPLATE Cetrimide agar

Heimplate, USP, EP, JP, plate diam. 90 mm, ICR Settle Plate for long Incubation, suitable for microbiology

Synonym(s):

Pseudomonas Selective Agar

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$73.60

$73.60


Available to ship on31 March 2025Details


Request more information

Select a Size

Change View
20 EA
$73.60

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

$73.60


Available to ship on31 March 2025Details


Request more information

Agency

EP
JP
USP

Quality Level

sterility

sterile

form

plate (ready-to-use)

feature

ready-to-use
ICR Settle Plate for long Incubation

packaging

pack of 20-120 plates

manufacturer/tradename

Heimplate

technique(s)

microbiological culture: suitable

plate diam.

90 mm

plate filling volume

30 mL

pH

7.2 (20 °C in H2O)

application(s)

microbiology
pharmaceutical
surface monitoring

storage temp.

15-25°C

suitability

Pseudomonas spp.

General description

Cetrimide Agar is suitable for selective isolation of Pseudomonas aeruginosa. The use of cetrimide (cetyltrimethylammonium bromide) largely inhibits the growth of the accompanying microbial flora. A concentration of 0.3 g/l of cetrimide inhibits the growth of accompanying organisms satisfactorily and minimizes interference with the growth of P. aeruginosa. The pigment production of P. aeruginosa is not inhibited when grown on this medium. P. aeruginosa can produce different pigments in different combinations. More than 90% of all strains produce the bluish, non-fluorescent pyocyanin, as well as the yellowish, fluorescent pyoverdin (fluorescein). Each settle plate is of 90 mm in diameter, and the packs are single-bagged in transparent, hydrogen peroxide impermeable sleeves (non-irradiated).

Application

Cetrimide Agar - LI is designed for selective isolation of Pseudomonas aeruginosa in non-sterile pharmaceutical products.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Questions

Reviews

Active Filters

  1. Prague
    • Review 1
    • Votes 0
    1 out of 5 stars.

    Mistake in Tech DS

    Pls, check and change TechDS.
    HEIMPLATE - what does it mean? Is it any new trade mark / brand name instead of LI-plate?

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for taking the time to leave a review! Per C25578469: HEIMPLATE is a trademark. The change of name from some LI plates to HEIMPLATE was a marketing decision to have the portfolio introduce a trademark. Other than the marketing, nothing has changed about this product. The documentation is currently being updated and will be available soon. If you have any other questions about this product, please contact us by visiting https://www.sigmaaldrich.com/support/customer-support and submit a Report Product Issue ticket.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service