Skip to Content
Merck
All Photos(1)

Key Documents

1.18322

Supelco

Chloride Test Refill pack

for use with MCOLORTEST®

Synonym(s):

Chloride Visual Test Kit Refills

Sign Into View Organizational & Contract Pricing

Select a Size

200 TESTS
$323.00

$323.00


Estimated to ship on02 April 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
200 TESTS
$323.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$323.00


Estimated to ship on02 April 2025


Request a Bulk OrderRequest more information

feature

refill

Quality Level

specific analyte(s)

chloride

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

When the reagents of a MQuant® test (formally MColortest®) are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of test kits, making it a particularly economical and ecological system.
This refill pack contains all the reagents for the MQuant® test kits 1.14753 an 1.14401 for the colometric evaluation of chlorine.
for 114753, 114401 MQuant®

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service