Skip to Content
Merck
All Photos(1)

Key Documents

1.14753

Supelco

Chloride Test Kit, colorimetric

3-300 mg/L (Cl⁻), MQuant®

Synonym(s):

Chloride Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

200 TESTS
$567.00

$567.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
200 TESTS
$567.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$567.00


Estimated to ship on29 May 2025


Request a Bulk OrderRequest more information

Product Name

Chloride Test, colorimetric, 3-300 mg/L (Cl-), for use with MCOLORTEST®

specific analyte(s)

chloride

Quality Level

measuring range

3-300 mg/L (Cl-)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Chloride is found in the earth´s crust and can therefore also be found in all natural waters at some concentration. This basic parameter of water analysis can be checked using the MQuant® Chloride test (formally MColortest®).

Method
Chloride ions react with mercury(II) thiocyanate to form slightly dissociated mercury(II) chloride. The thiocyanate released in the process in turn reacts with iron(III) ions to form red iron(III) thiocyanate. The chloride concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk. The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation.
The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments. Using the refill pack 1.18322 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.

This test can be used in
  • Groundwater, surface water, and seawater (after dilution)
  • Drinking water and mineral water
  • Waters from aquaculture
  • Boiler and boiler feed water, cooling water
  • Industrial water
  • Wastewater and percolating water
  • Process water
  • Swimming-pool water
  • Food after appropriate sample pretreatment
  • Soils after appropriate sample pretreatment
Method: colorimetric with color-disk comparator 3 - 6 - 10 - 18 - 30 - 60 - 100 - 180 - 300 mg/l Cl MQuant®

Application

  • PREFUL MRI for Monitoring Perfusion and Ventilation Changes after Elexacaftor-Tezacaftor-Ivacaftor Therapy for Cystic Fibrosis: A Feasibility Study.: This study demonstrates the use of PREFUL MRI to monitor changes in perfusion and ventilation in cystic fibrosis patients post-therapy, showcasing its potential for non-invasive assessment in clinical settings (Dohna et al., 2024).
  • Wearable sweat chloride sensors: materials, fabrication and their applications.: Reviews advances in materials and fabrication techniques for wearable sweat chloride sensors, discussing their significant potential for real-time health monitoring and diagnostics (Patel et al., 2024).
  • Initial characteristics of cystic fibrosis in Algeria: Description of 34 pediatric cases.: Provides an overview of the clinical presentation and initial characteristics of cystic fibrosis in a pediatric Algerian cohort, contributing to the regional understanding of this genetic disorder (Bendoukha et al., 2024).

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service