Skip to Content
Merck
All Photos(1)

Documents

1.10079

Supelco

Chloride Test Strips, colorimetric

500-3000 mg/L (Cl⁻), MQuant®

Synonym(s):

Chloride chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Chloride Test, colorimetric, 500-3000 mg/L (Cl-), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

chloride

measuring range

500-3000 mg/L (Cl-)

detection method

colorimetric

storage temp.

2-8°C

General description

Method: Colorimetric, with test strips, 500 - 1000 - 1500 - 2000 - ≥ 3000 mg/L (Cl-), MQuant®

The MQuant® Chloride test strips are suitable for measuring chloride ions in all natural waters. Their concentration depends on geological factors and the general local situation. Chloride ions react with silver ions, decolorizing red-brown silver chromate. The chloride concentration is measured semi-quantitatively by visual comparison of the reaction zones of the test strip with the color rows of a color scale. Especially prior to performing a COD (chemical oxygen demand) test it is recommended to measure the Chloride level of the sample as this is a mayor interference. Pre-testing samples with MQuant® Chloride test strips allows to choose the correct dilution factor and measurement range for COD testing which saves time and resources.

Linkage

Replaces: 10079-1; 10079

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service