Skip to Content
Merck
All Photos(5)

Key Documents

1.00997

Supelco

1-Propanol

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

1-Propanol, n-Propyl alcohol, Propan-1-ol

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$133.00
2.5 L
$276.00
4 L
$442.00
25 L
$1,600.00

$133.00


Available to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
$133.00
2.5 L
$276.00
4 L
$442.00
25 L
$1,600.00

About This Item

Empirical Formula (Hill Notation):
C3H8O
CAS Number:
Molecular Weight:
60.10
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-746-9
NACRES:
NB.21
grade:
ACS reagent
for analytical purposes
bp:
96.5-98 °C/1013 hPa
vapor pressure:
19 hPa ( 20 °C)

$133.00


Available to ship on01 April 2025Details


Request a Bulk Order

grade

ACS reagent
for analytical purposes

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

19 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

360 °C

expl. lim.

2.1-19.2 % (v/v)

dilution

(for analytical testing)

impurities

≤0.0002 meq/g Alkalinity
≤0.0004 meq/g Acidity
≤0.01% Acetone (GC)
≤0.01% Ethanol (GC)
≤0.01% Methanol (GC)
≤0.03% Aldehydes and ketones (as C3H6O)
≤0.05% 2-Propanol (GC)
≤0.05% Water

evapn. residue

≤0.001%

color

clearAPHA: ≤10

pH

7 (20 °C, 200 g/L in H2O)

bp

96.5-98 °C/1013 hPa

mp

-127 °C

transition temp

flash point 15 °C

density

0.80 g/cm3 at 20 °C

cation traces

Al: ≤0.00005%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

storage temp.

2-30°C

SMILES string

OCCC

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application


  • A novel high-throughput analytical method to quantify microplastics in water by flow cytometry.: In this paper water is used to wash the Natural aquarium silica sand and also to prepare the model for the removal of microplastics. (Li et al., 2023).


Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

  1. Can the product be used in GC?

    1 answer
    1. Yes, this product may be used for Gas Chromatography. The purity of this product is determined by GC.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service