Skip to Content
Merck
All Photos(1)

Key Documents

1.16896

Supelco

Chlorine Test Strips, reflectometric

0.5-10.0 mg/L (Cl₂), REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

Chlorine chemical test strips, Chlorine test strip for instrument reader

Sign Into View Organizational & Contract Pricing

Select a Size

50 TESTS
$172.00

$172.00


Available to ship on07 April 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
50 TESTS
$172.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$172.00


Available to ship on07 April 2025Details


Request a Bulk OrderRequest more information

Product Name

Chlorine Test, reflectometric, 0.5-10.0 mg/L (Cl2), for use with REFLECTOQUANT®

description

quantitative

Quality Level

product line

Reflectoquant®

specific analyte(s)

chlorine

reaction suitability

reaction type: Redox Reactions

measuring range

0.5-10.0 mg/L (Cl2)

compatibility

RQflex® 20
for use with REFLECTOQUANT®

detection method

reflectometric

storage temp.

2-8°C

General description

Method: reflectometric with test strips and reagent 0.5 - 10.0 mg/l Cl₂ Reflectoquant®
The Reflectoquant Chlorine test includes test strips and all necessary additional reagents and is a rapid exploratory test for free chlorine and intended primarily for checking the use of chlorinated disinfectants. Chlorine containing disinfectants are most widely used. They are applied particularly to disinfect drinking water and swimming pool water. On the test strip, chlorine oxidizes an organic compound to a violet dye that is determined reflectometrically on the RQflex® 20. For pre-testing a sample in an economic and quick way to estimate the chlorine concentration and a possible dilution factor prior to using this quantitative method we recommend our semi-quantitative MQuant test strips for chlorine. Further quantitative determination methods can be found with our Spectroquant tests.

Analysis Note

function control: conforms
Assay
- standard 1:
1.8 - 2.2 mg/l
- standard 2: 4.5 - 5.5 mg/l
- standard 3: 7.2 - 8.8 mg/l

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about disinfection control and how to select the right disinfectant among the various options available in its methods and instrumentation based on photometry and reflectometry.

Protocols

Preparation of a standard solution for total chlorine.

AQA Standard for free chlorine - Preparation of a standard solution for free chlorine

Preparation of a standard solution for free Chlorine according to DIN EN ISO 7393

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service