Skip to Content
Merck
All Photos(1)

Key Documents

1.18389

Supelco

Sulfate Test Kit, colorimetric

25-300 mg/L (SO₄²⁻), MQuant®

Sign Into View Organizational & Contract Pricing

Select a Size

75 TESTS
$893.00

$893.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
75 TESTS
$893.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$893.00


Estimated to ship on28 May 2025


Request a Bulk OrderRequest more information

Product Name

Sulfate Test, colorimetric, 25-300 mg/L (SO42-), for use with MCOLORTEST®

specific analyte(s)

sulfate

Quality Level

measuring range

25-300 mg/L (SO42-)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color-disk comparator 25 - 50 - 75 - 100 - 130 - 160 - 190 - 240 - 300 mg/l SO MQuant®
The MQuant™ Sulfate test (formally MColortest™) is used for the determination of sulfate in:


  • Groundwater and surface water
  • Drinking water and mineral water
  • Wastewater

    This test is not suited for seawater.
    Method
    Sulfate ions react with barium iodate, liberating iodate ions in the process. These oxidize tannin to a brown-red compound. The sulfate concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color disk The test evaluates the color reaction based on the transmitted light method. So even turbid and colored water samples can be analyzed without further preparation. The color disk is made of very durable, lightfast plastic, thus suitable for industrial areas and wet environments.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Liq. 2 - Repr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

147.2 °F

Flash Point(C)

64 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service