Skip to Content
Merck
All Photos(1)

Key Documents

1.11152

Supelco

Oxygen Test Refill pack

for use with MCOLORTEST®

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$273.00

$273.00


Estimated to ship on09 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
100 TESTS
$273.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$273.00


Estimated to ship on09 May 2025


Request a Bulk OrderRequest more information

feature

refill

Quality Level

analyte chemical class(es)

oxygenates

compatibility

for use with MCOLORTEST®

detection method

titrimetric

storage temp.

15-25°C

General description

When the reagents of a MQuant® test (formally MColortest®) are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of test kits, making it a particularly economical and ecological system.
This refill pack contains all reagents for the MQuant® test kit 1.11107 for the titrimetric evaluation of oxygen.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT RE 2 - STOT RE 2 Oral

Target Organs

Brain, Thyroid

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service