Skip to Content
Merck
All Photos(1)

Documents

1.11151

Supelco

Compact Laboratory for Water Testing

for use with MCOLORTEST®

Synonym(s):

Portable Water Testing Laboratory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

analyte chemical class(es)

oxygenates

Quality Level

specific analyte(s)

ammonium, calcium, magnesium, nitrate, nitrite, phosphate

compatibility

for use with MCOLORTEST®

detection method

colorimetric
titrimetric

storage temp.

15-25°C

General description

The MQuant® Compact Laboratory for Water Testing (formally MColortest®) contains all the reagents and auxiliaries necessary for the determination of: ammonium, carbonate hardness (acidbinding capacity), total hardness (and residual hardness), nitrate, nitrite, pH, phosphate, oxygen and oxygen consumption, and temperature.
The Compact Laboratory is suited for numerous applications, for example for monitoring the quality of waters, for supporting analyses in connection with the biological assessment of waters, and also in the academic and teaching fields. The tests themselves are based on colorimetric and titrimetric methods. On the enclosed color cards to be used for the colorimetric tests brief illustrated instructions are provided.

Analysis Note

Application test: conforms

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Muta. 2 - Repr. 1B - Skin Corr. 1A - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service