Skip to Content
Merck
All Photos(2)

Key Documents

1.08047

Supelco

Total Hardness Test Kit, titrimetric

titrimetric, for use with MQUANT®

Synonym(s):

Total Hardness Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

300 TESTS
$169.00

$169.00


Estimated to ship on01 June 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
300 TESTS
$169.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$169.00


Estimated to ship on01 June 2025


Request a Bulk OrderRequest more information

Product Name

Total Hardness Test, titrimetric, for use with MQUANT®

specific analyte(s)

calcium, magnesium

Quality Level

compatibility

for use with MQUANT®

detection method

titrimetric

storage temp.

15-25°C

General description

Method: titrimetric with titration pipette MQuant®
This MQuant® Total Hardness test (formally MColortest®) is suitable for the titrimetric determination using a graduated syringe. It can be used in:
  • Groundwater and surface water, seawater
  • Drinking water and mineral water
  • Waters from aquaculture
  • Swimming-pool water
  • Boiler water

Method
Calcium and magnesium ions react with an indicator to form a red complex compound. The indicator is released from this compound by titration with a solution of ethylenedinitrilotetraacetic acid disodium salt dihydrate (Titriplex® III). At the titration end-point the color changes to green. The total hardness is determined from the consumption of titration solution.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service