Skip to Content
Merck

Skip To

1.10012

Chromate Test Strips, colorimetric

greener alternative

3-100 mg/L (CrO₄²⁻), MQuant®

Synonym(s):

Chromate chemical test strips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 tests

Available to ship TODAYfromTruganina

$190.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$190.00


Available to ship TODAYDetails


Request a Bulk OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Chromate Test, colorimetric, 3-100 mg/L (CrO42-), MQuant®

description

semi-quantitative

Quality Segment

product line

MQuant®

specific analyte(s)

chromate

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

3-100 mg/L (CrO42-)

application(s)

electroplating
wastewater

detection method

colorimetric

greener alternative category

storage temp.

15-25°C

General description

Method: Colorimetric, with test strips and reagent, 3 - 10 - 30 - 100 mg/L (CrO42-), MQuant®

The MQuant® Chromate test includes test strips and all necessary addidtional reagents. It can be used to monitor waste water discharged from electroplating and pickling plants, in cement and for rapidly and reliably determining of chromium(VI) salts. In weakly phosphoric solution chromium(VI) ions react with diphenylcarbazide to form chromium(III) and diphenylcarbazone, which form a red-violet complex. The concentration of chromate is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the chromate concentration and a possible dilution factor prior to using a quantitative method such as a Spectroquant® portfolio.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Application


  • Environmental Monitoring: Chromate tests are essential for monitoring chromium(VI) levels in water and soil. These tests help in assessing environmental pollution caused by industrial discharge and are crucial for ensuring compliance with environmental regulations. Recent research has focused on the simultaneous extraction of phosphate and chromate using nano mixed (Al-Fe-Zr) oxide adsorbents, demonstrating enhanced sensitivity and accuracy in detecting chromate contamination in water (Pala et al., International Journal of Environmental Analytical Chemistry, 2021).



Other Notes

Replaces: 10012-1

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.100441.100021.10049
detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

specific analyte(s)

chromate

specific analyte(s)

cyanide

specific analyte(s)

cobalt

specific analyte(s)

molybdenum

product line

MQuant®

product line

MQuant®

product line

MQuant®

product line

MQuant®

application(s)

electroplating
wastewater

application(s)

electroplating
wastewater

application(s)

electroplating
wastewater

application(s)

boiler and cooling water

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

measuring range

3-100 mg/L (CrO42-)

measuring range

-

measuring range

10-1000 mg/L (Co2+)

measuring range

5-250 mg/L (Mo)


pictograms

Corrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

wgk

WGK 1

hcodes

Storage Class

8A - Combustible corrosive hazardous materials



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
110012000104022536120867

Questions

  1. Is there a “how to use” video, that shows all the stages of treatment you can use the strip?

    1 answer
    1. There is no "how to use" video for this product. A product bulletin with procedure information is available at https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/168/793/110012ug-ms.pdf

      Helpful?

Reviews

No rating value

Active Filters