Skip to Content
Merck
All Photos(1)

Key Documents

1.09584

Supelco

pH-indicator strips pH 2.0 - 9.0

non-bleeding, colorimetric, pH range 2.0-9.0, graduations and accuracy: 0.5 pH unit, MQuant®

Synonym(s):

pH dip stick pH 2.0 - 9.0, pH test strips pH 2.0 - 9.0

Sign Into View Organizational & Contract Pricing

Select a Size

100 STRIPS
$52.50

$52.50


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
100 STRIPS
$52.50

About This Item

UNSPSC Code:
41113034
NACRES:
NB.35

$52.50


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

Product Name

pH-indicator strips pH 2.0 - 9.0, non-bleeding, colorimetric, pH range 2.0-9.0, graduations and accuracy accuracy: 0.5 pH unit, MQuant®

product line

MQuant®

Quality Level

feature

non-bleeding

graduations and accuracy measuring range

accuracy: 0.5 pH unit

pH range

2.0-9.0

detection method

colorimetric

storage temp.

2-30°C

General description

The MQuant pH tests are suited for all media in environmental analysis and in industrial in-process controls. Our special method of manufacture gives the pH indicator strips decisive advantages over the indicator paper: since the indicator does not bleed, the strip can be left in the measurement medium without contaminating it. Using these test strips is fairly easy, the pH value is measured semiquantitatively by visual comparison of the three reaction zones on the test strip with the fields of the color scale, perfectly matching the reaction color. The clever Safetyedge boxes allow for safe storage and convenient withdrawal of the test strips. For tests for other parameters that require a specific pH range of the sample (e.g. Spectroquant, see instructions of use), we recommend the use of MQuant pH test strips as pre-test.
non-bleeding 2.0 - 2.5 - 3.0 - 3.5 - 4.0 - 4.5 - 5.0 - 5.5 - 6.0 - 6.5 - 7.0 - 7.5 - 8.0 - 8.5 - 9.0 MQuant®

Linkage

Replaces: 9578-1; 9578-3; 9578

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Quantify selenium in water using this photometric test and a clear, filtered sample to measure the results at 445 nm.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service