Skip to Content
Merck
All Photos(3)

Key Documents

1.09502

Supelco

pH-indicator strips pH 2.0 - 9.0

non-bleeding, colorimetric, pH range 2.0-9.0, for use in turbid solutions (suspensions), graduations and accuracy: 1 pH unit, MQuant®

Synonym(s):

pH dip stick pH 2.0 - 9.0, pH test strips pH 2.0 - 9.0

Sign Into View Organizational & Contract Pricing

Select a Size

100 STRIPS
$28.90

$28.90


Available to ship on02 April 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
100 STRIPS
$28.90

About This Item

UNSPSC Code:
41113034
NACRES:
NB.35

$28.90


Available to ship on02 April 2025Details


Request a Bulk OrderRequest more information

Product Name

pH-indicator strips pH 2.0 - 9.0, non-bleeding, colorimetric, pH range 2.0-9.0, graduations and accuracy accuracy: 1 pH unit, MQuant®

product line

MQuant®

Quality Level

feature

non-bleeding

graduations and accuracy measuring range

accuracy: 1 pH unit

pH range

2.0-9.0

detection method

colorimetric

storage temp.

2-30°C

General description

Special indicator test strip for the pH measurement also in turbid samples. The MQuant pH tests are suited for all media in environmental analysis and in industrial in-process controls. Our special method of manufacture gives the pH indicator strips decisive advantages over the indicator paper: since the indicator does not bleed, the strip can be left in the measurement medium without contaminating it. Using these test strips is fairly easy, the pH value is measured semiquantitatively by visual comparison of the three reaction zones on the test strip with the fields of the color scale, perfectly matching the reaction color. For tests for other parameters that require a specific pH range of the sample (e.g. Spectroquant, see instructions of use), we recommend the use of MQuant pH test strips as pre-test.
for pH measurements in turbid solutions (suspensions) non-bleeding pH 2 - 3 - 4 - 5 - 6 - 7 - 8 - 9 MColorpHast

Legal Information

MCOLORPHAST is a trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service