Skip to Content
Merck
All Photos(1)

Key Documents

1.09540

Supelco

pH-indicator strips pH 0 - 2.5

non-bleeding, colorimetric, pH range 0-2.5, MQuant®

Synonym(s):

pH dip stick pH 0 - 2.5, pH test strips pH 0 - 2.5

Sign Into View Organizational & Contract Pricing

Select a Size

100 STRIPS
$52.50

$52.50


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
100 STRIPS
$52.50

About This Item

UNSPSC Code:
41113034
NACRES:
NB.35

$52.50


Estimated to ship on30 May 2025


Request a Bulk OrderRequest more information

product line

MQuant®

Quality Level

feature

non-bleeding

pH range

0-2.5

detection method

colorimetric

storage temp.

2-30°C

General description

The MQuant® pH tests are suited for all media in environmental analysis and in industrial in-process controls. Our special method of manufacture gives the pH indicator strips decisive advantages over the indicator paper: since the indicator does not bleed, the strip can be left in the measurement medium without contaminating it. Using these test strips is fairly easy, the pH value is measured semiquantitatively by visual comparison of the reaction zone on the test strip with the fields of the color scale, perfectly matching the reaction color. The clever Safetyedge boxes allow for safe storage and convenient withdrawal of the test strips. For tests for other parameters that require a specific pH range of the sample (e.g. Spectroquant®, see instructions of use), we recommend the use of MQuant® pH test strips as pre-test.
non-bleeding pH 0 - 0.5 - 1.0 - 1.3 - 1.6 - 1.9 - 2.2 - 2.5 MQuant®

Linkage

Replaces: 9580-1; 9580-3; 9580

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service