Skip to Content
Merck
All Photos(1)

Key Documents

1.03025

Supelco

Bromocresol purple

pH indicator, ACS, Reag. Ph Eur

Synonym(s):

Bromocresol purple, 5′,5′′-Dibromo-o-cresolsulfonphthalein

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$135.00
25 G
$337.00

$135.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$135.00
25 G
$337.00

About This Item

Empirical Formula (Hill Notation):
C21H16Br2O5S
CAS Number:
Molecular Weight:
540.22
MDL number:
UNSPSC Code:
12171502
EC Index Number:
204-087-8
NACRES:
NA.21

$135.00


Estimated to ship on28 May 2025


Request a Bulk Order

Product Name

Bromocresol purple, indicator ACS,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

loss

≤1% loss on drying, 110°C

visual transition interval

5.2-6.8, greenish yellow to blue-violet

mp

242 °C

bulk density

515 kg/m3

storage temp.

2-30°C

SMILES string

[S]1(=O)(=O)OC(c4c1cccc4)(c3cc(c(c(c3)C)O)Br)c2cc(c(c(c2)C)O)Br

InChI

1S/C21H16Br2O5S/c1-11-7-13(9-16(22)19(11)24)21(14-8-12(2)20(25)17(23)10-14)15-5-3-4-6-18(15)29(26,27)28-21/h3-10,24-25H,1-2H3

InChI key

ABIUHPWEYMSGSR-UHFFFAOYSA-N

Analysis Note

Transition range: pH 5.2 - pH 6.8greenish yellow - blue violet
Appearance of solution: passes test
Loss on drying (110 °C): ≤ 1
Transition range (according to ACS): passes test
Sensitivity test: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service