Skip to Content
Merck
All Photos(1)

Key Documents

1.03026

Supelco

Bromothymol blue

pH indicator, ACS, Reag. Ph Eur

Synonym(s):

Bromothymol blue, 3′,3′′-Dibromothymolsulfonphthalein, BTB

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$179.00
25 G
$449.00

$179.00


Available to ship on01 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$179.00
25 G
$449.00

About This Item

Empirical Formula (Hill Notation):
C27H28Br2O5S
CAS Number:
Molecular Weight:
624.38
MDL number:
UNSPSC Code:
12171502
EC Index Number:
200-971-2
NACRES:
NB.21

$179.00


Available to ship on01 April 2025Details


Request a Bulk Order

Product Name

Bromothymol blue, indicator ACS,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

loss

≤3% loss on drying, 110°C

visual transition interval

5.8-7.6, yellow to blue

bulk density

450 kg/m3

storage temp.

2-30°C

SMILES string

[S]1(=O)(=O)OC(c4c1cccc4)(c3c(c(c(c(c3)C(C)C)O)Br)C)c2c(c(c(c(c2)C(C)C)O)Br)C

InChI

1S/C27H28Br2O5S/c1-13(2)17-11-20(15(5)23(28)25(17)30)27(19-9-7-8-10-22(19)35(32,33)34-27)21-12-18(14(3)4)26(31)24(29)16(21)6/h7-14,30-31H,1-6H3

InChI key

NUHCTOLBWMJMLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Novel pH-responsive indicator films based on bromothymol blue-anchored chitin for shrimp freshness monitoring: This study presents innovative pH-sensitive films that incorporate bromothymol blue into chitin, designed for tracking shrimp spoilage. The development and testing of these films in real-time scenarios offer a viable method for monitoring seafood freshness, ensuring quality control in food distribution chains (Yang et al., 2023).

Analysis Note

Transition range: pH 5.8 - pH 7.6yellow - blue
Appearance of solution: passes test
Loss on drying (110 °C): ≤ 3
Transition range (according to ACS): passes test
Sensitivity test: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service