Skip to Content
Merck
All Photos(2)

Key Documents

05-1064

Sigma-Aldrich

Anti-Phospho-Stat1 (Tyr701) Antibody, clone 5C9.2

clone 5C9.2, from mouse

Synonym(s):

Transcription factor ISGF-3 components p91/p84, signal transducer and activator of transcription 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$567.00

$567.00


Estimated to ship on30 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1520

Request a Bulk Order

Select a Size

Change View
100 μG
$567.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$567.00


Estimated to ship on30 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1520

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5C9.2, monoclonal

species reactivity

rat, mouse, human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr701)

Gene Information

human ... STAT1(6772)
mouse ... Stat1(20846)

General description

STATs (Signal-Transducing Activators of Transcription) are the principle substrates of JAK kinases, and mediators of cytokine signaling. STAT proteins have SH2 domains, and once phosphorylated by JAK kinases, STATs dimerize in a head-to-tail fashion, through their SH2 domains. STAT1 is a critical mediator of IFN signaling, STAT4 is specific to IL-12, STAT6 is activated primarily by IL-4 and IL-13, while STAT5 A and B appear to be most important to the function of growth hormone and prolactin. STAT3 is involved in multiple signaling pathways, and mice deficient in it are not viable. In addition to tyrosine phosphorylation, many STAT proteins are targets of Ser/Thr kinases, and those phosphorylation events potentiate STAT-induced transcription. The phosphorylation state of STATs can be monitored by phosphorylation state-specific antibodies.

Specificity

This antibody recognizes phosphorylated STAT1 at tyrosine 701.

Immunogen

Epitope: Phosphorylated Tyr701
KLH-conjugated linear peptide corresponding to the region around phosphorylated tyrosine 701.

Application

Anti-Phospho-Stat1 (Tyr701) Antibody, clone 5C9.2 detects level of Phospho-Stat1 (Tyr701) & has been published & validated for use in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Routinely evaluated by western blot on EGF treated and untreated A431 cell lysate.

Western Blot Analysis: 0.5-2 µg/mL of this lot was used to detect STAT1 phosphorylated at Tyr701 in EGF treated and untreated A431 cell lysates.

Target description

88-92 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing PBS and 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
EGF treated and untreated A431 cell lysates, PDGF treated and untreated NIH/3T3 cell lysates, pervanadate treated and untreated PC12 cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Darnell
Science (New York, N.Y.), 277(5332), 1630-1635 (1997-09-12)
STATs (signal transducers and activators of transcription) are a family of latent cytoplasmic proteins that are activated to participate in gene control when cells encounter various extracellular polypeptides. Biochemical and molecular genetic explorations have defined a single tyrosine phosphorylation site

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service