Skip to Content
Merck
All Photos(1)

Key Documents

AB2034

Sigma-Aldrich

Anti-Anti-Laminin (LAMA1) antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

Anti-LAMA, Anti-PTBHS, Anti-S-LAM-alpha

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$785.00

$785.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$785.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$785.00


Estimated to ship on30 May 2025


Request a Bulk Order

Product Name

Anti-Laminin Antibody, Chemicon®, from rabbit

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
radioimmunoassay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Lama1(16772)

Specificity

Laminin extracted and purified from mouse tumor tissues. Antibody shows less than 0.1% reactivity with mouse collagen Types I, II, and IV; rat collagen Types I, III, and V; rat and human elastin; mouse fibronectin and laminin by radioimmunoassay.

Immunogen

Laminin extracted and purified from EHC mouse tumor tissues.

Application

Anti-Laminin Antibody detects level of Laminin & has been published & validated for use in ELISA, RIA, IH.
IFA (with fluorescein anti-rabbit IgG conjugate) - use diluted at ≥ 1:80 on frozen mouse tissues (skin, liver, etc.).
IHC (paraffin) - use diluted at ≥ 1:500 on fixed, paraffin-embedded mouse tissues (skin, liver, etc). (immunoperoxidase technique with the Envision / Dako kit);
ELISA - use diluted at ≥ 1:200 (OD ≥ 500).
Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

The purified antibody is supplied in a buffer containing a mixture of 0.1M Citrate, 0.1M potassium phosphate, at a pH of 7.2-7.4 and 10µl/ml of antibiotics and antimycotics.

Storage and Stability

Maintain frozen at -20°C for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pei-Lin Shao et al.
PloS one, 11(6), e0156699-e0156699 (2016-06-02)
Micro-plasma is a possible alternative treatment for wound management. The effect of micro-plasma on wound healing depends on its composition and temperature. The authors previously developed a capillary-tube-based micro-plasma system that can generate micro-plasma with a high nitric oxide-containing species
Lifei Hou et al.
Blood advances, 4(24), 6086-6097 (2020-12-23)
β2 integrins are well-known leukocyte adhesion molecules consisting of 4 members: CD11a-d. Their known biological functions range widely from leukocyte recruitment, phagocytosis, to immunological synapse formation, but the studies have been primarily focused on CD11a and CD11b. CD11c is 1
Xin Wu et al.
Frontiers in nutrition, 9, 897821-897821 (2022-09-27)
Aging-related muscle loss is a hallmark of aging and is the cause of some negative outcomes. An optimized diet and supplements have a positive effect in slowing down the process of muscle loss. D-galactose(d-gal) has been used widely to develop
Zitao Liu et al.
Biology of reproduction, 100(1), 217-226 (2018-08-08)
Heparin-binding epidermal growth factor (EGF)-like growth factor (HBEGF) is expressed in the embryo and uterus at the implantation site, stimulating trophoblast invasive activity essential for placentation. The effect of extraembryonic HBEGF deficiency on placental development was investigated by breeding mice
Chan Ho Hwang et al.
Development (Cambridge, England), 146(4) (2019-02-17)
The semicircular canals of the mammalian inner ear are derived from epithelial pouches in which epithelial cells in the central region of each pouch undergo resorption, leaving behind the region at the rim to form a tube-shaped canal. Lack of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service