Skip to Content
Merck
All Photos(1)

Key Documents

890830C

Avanti

16:0 DAP

Avanti Research - A Croda Brand 890830C

Synonym(s):

1,2-dipalmitoyl-3-dimethylammonium-propane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H73NO4
CAS Number:
Molecular Weight:
595.98
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (890830C-25mg)
pkg of 2 × 4 mL (890830C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890830C

concentration

10 mg/mL (890830C-25mg)
25 mg/mL (890830C-200mg)

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(CN(C)C)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

Application

1,2-dipalmitoyl-3-dimethylammonium-propane (16:0 DAP) is suitable for the preparation of miRNA encapsulated liposomes.

Biochem/physiol Actions

1,2-dipalmitoyl-3-dimethylammonium-propane (16:0 DAP) is a cationic lipid and serves as a pH-sensitive transfection reagent.

Packaging

5 mL Clear Glass Sealed Ampule (890830C-200mg)
5 mL Clear Glass Sealed Ampule (890830C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moran Amit et al.
Nature, 578(7795), 449-454 (2020-02-14)
The solid tumour microenvironment includes nerve fibres that arise from the peripheral nervous system1,2. Recent work indicates that newly formed adrenergic nerve fibres promote tumour growth, but the origin of these nerves and the mechanism of their inception are unknown1,3.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service