Skip to Content
Merck
All Photos(1)

Key Documents

PBVK02510

Millipore

PES Membrane Filter, 500 kDa NMWCO

Biomax®, filter diam. 25 mm

Synonym(s):

Biomax PES ultrafiltration membrane discs, Ultrafiltration Discs, 500 kDa NMW

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$244.00

$244.00


Check Cart for Availability


Select a Size

Change View
10 EA
$244.00

About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602
NACRES:
NB.24

$244.00


Check Cart for Availability

material

polyethersulfone membrane

Quality Level

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Biomax®

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

diam.

25 mm

filter diam.

25 mm

filtration area

4.1 cm2

pore size

500 kDa NMWCO

shipped in

ambient

Application

500 kDa NMWL, Biomax polyethersulfone, 25 mm diameter

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service